Merck
  • Crystal structure of CelM2, a bifunctional glucanase-xylanase protein from a metagenome library.
Crystal structure of CelM2, a bifunctional glucanase-xylanase protein from a metagenome library.
Biochemical and biophysical research communications (2009-04-07)
Ki Hyun Nam, Soo-Jin Kim, Kwang Yeon Hwang
ABSTRACT
Degradation of polysaccharides by cellulases and xylanases plays an important role in the carbon cycle, but only occurs in plant cell walls, a few bacteria and some animals. This process is also critical in processes such as biomass degradation and fuel production in the conversion cycles of cellulosic biomass. The enzyme CelM2 is bifunctional, because it is able to effectively hydrolyze barley glucan and xylan. Here, we show the crystal structure of the bifunctional enzyme CelM2, isolated from a metagenome library, and describe the sequence information and structure of its two domains. We believe that CelM2 is attractive as an industrial enzyme and that the structural results presented herein provide insights that are relevant to the genetic engineering of multifunctional enzymes.
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 400-800 units/mg protein
Sigma-Aldrich
Sustine 470
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 10-25 units/mg solid
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 100-250 units/mg protein