Merck
[Jasmonate biosynthesis--the latest discoveries].
Postepy biochemii (2012-12-12)
Emilia Wilmowicz, Kamil Frankowski, Magdalena Sidłowska, Agata Kućko, Jacek Kesy, Adam Gasiorowski, Paulina Glazińska, Jan Kopcewicz
ABSTRACT
Jasmonates are plant hormones involved in many growth and development processes. They also participate in plant defense responses. Current progress in the study on biosynthesis and signaling of jasmonates has contributed to the understanding of the mechanisms regulating concentration of these hormones in the cell. Sustaining a proper level of jasmonates allow the plant to respond appropriately to changing conditions. It is possible due to the large number of enzymes and genes involved in biosynthesis of these hormones as well as multilevel control of their expression.
Sigma-Aldrich
Lipoxidase from Glycine max (soybean), Type V, ammonium sulfate suspension, 500,000-1,000,000 units/mg protein
Sigma-Aldrich
Lipoxidase from Glycine max (soybean), Type I-B, lyophilized powder, ≥50,000 units/mg solid