Merck
  • Evaluation of an electrochemical method for the analysis of enzymatic inhibition reactions.
Evaluation of an electrochemical method for the analysis of enzymatic inhibition reactions.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2013-01-11)
Kohei Uematsu, Hajime Katano
ABSTRACT
An approximate equation for bioelectrocatalitic current was applied to an inhibition reaction analysis of bilirubin oxidase by anion (Cl(-), SCN(-), and F(-)) in order to assess the possibility of the electrochemical method for the analysis of enzymatic inhibition reactions. The approximate equation can be transformed into the Michaelis-Menten form, so that the bioelectrocatalitic current can be analyzed by the usual graphical manner, that is, Lineweaver-Burk, Hanes-Woolf, Dixon and Cornish-Bowden plots, if the rate of inhibition reaction is described as a simple Michaelis-Menten form. From the electrochemical assay, it was found that the inhibitor of Cl(-) and SCN(-) anions exhibit non-competitive inhibition while that of F(-) exhibits competitive inhibition, and their inhibition constants were 220, 45, and 22 mM, respectively. The results were essentially similar to those obtained from the conventional spectrophotometric assay.
Sigma-Aldrich
Bilirubin Oxidase from Myrothecium verrucaria, lyophilized powder, 15-65 units/mg protein
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium thiocyanate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium thiocyanate solution, BioUltra, 8 M in H2O
Sigma-Aldrich
Sodium thiocyanate, reagent grade, 98-102% (titration)
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium thiocyanate, ACS reagent, ≥98.0%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%