Merck
  • Effect of the steroid K-canrenoate on hsp70 expression and tissue damage in transgenic Drosophila melanogaster (hsp70-lacZ) Bg9.
Effect of the steroid K-canrenoate on hsp70 expression and tissue damage in transgenic Drosophila melanogaster (hsp70-lacZ) Bg9.
Journal of insect science (Online) (2013-02-23)
Yasir Hasan Siddique, Gulshan Ara, Mohammad Afzal
ABSTRACT
In the present study the effect of 0.1, 0.2, 0.4, 0.8, and 1.0 µL/mL of the steroid K-canrenoate was evaluated in the third instar larvae of transgenic Drosophila melanogaster (hsp70-lacZ) Bg(9) for 6, 24, and 48 hours of duration. The treatment of 0.1, 0.2, and 0.4 µL/mL of K-canrenoate did not induce the activity of hsp70 significantly compared to the control. The treatments of 0.8 and 1.0 µL/mL of K-canrenoate not only caused tissue damage but also induced a significant increase in the expression of hsp70 for the different durations of exposure. The results of the present study suggest that the K-canrenoate at 0.8 and 1.0 µL/mL is cytotoxic and caused tissue damage in the third instar larvae of transgenic D. melanogaster (hsp70-lacZ) Bg(9).
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
β-Galactosidase from Escherichia coli, suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Sigma-Aldrich
β-Galactosidase from Escherichia coli, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
β-Galactosidase from bovine liver, Grade III, lyophilized powder, ≥0.15 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VI, lyophilized powder, ≥250 units/mg protein
Sigma-Aldrich
β-Galactosidase from bovine testes, ammonium sulfate suspension, 1.0-3.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
β-Galactosidase from Escherichia coli, aqueous glycerol suspension, ≥500 units/mg protein (biuret)
Sigma-Aldrich
Canrenoic acid potassium salt, powder
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
β-Galactosidase from Aspergillus oryzae, ≥8.0 units/mg solid