Merck
  • Screening and identification of newly isolated cellulose-degrading bacteria from the gut of xylophagous termite Microcerotermes diversus (Silvestri).
Screening and identification of newly isolated cellulose-degrading bacteria from the gut of xylophagous termite Microcerotermes diversus (Silvestri).
Mikrobiologiia (2013-04-25)
Z Pourramezan, G R Ghezelbash, B Romani, S Ziaei, A Hedayatkhah
ABSTRACT
The aim of the present study was to isolate and characterize the cellulose-degrading bacteria from the gut of the local termite, Microcerotermes diversus (Silvestri), inhabiting the Khuzestan province of Iran. The microorganisms capable of growing in the liquid medium containing cellulose as the only source of carbon were isolated and their cellulolytic activity on CMC-containing media was confirmed by the congo red clearing zone assay. The isolates were identified based on biochemical characteristics and the phylogenetic analysis of 16S rRNA gene fragments. The results of the present study show that three cellulose-degrading bacteria isolated from local termite guts belonged to the genera Acinetobacter, Pseudomonas and Staphylococcus and four cellulose-degrading bacteria belonged to Enterobacteriaceae and Bacillaceae families. Several isolates recovered from separate termite Microcerotermes diversus samples closely clustered in phylogenetic trees indicating high similarity and the abundance of particular cellulolytic strains. Bacillus B5B and Acinetobacter L9B hydrolyzed cellulose faster than the other isolates (with CMCase activity of 1.47 U/mL and 1.22 U/mL, respectively). The stability of CMCase produced by Bacillus B5B over a broad range of pH and high temperature indicated that the enzyme may be of great commercial value.
Sigma-Aldrich
Carboxymethylcellulose sodium salt, ultra high viscosity, highly purified
Supelco
Congo Red, analytical standard
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulase from Trichoderma reesei ATCC 26921, lyophilized powder, ≥1 unit/mg solid
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity
Sigma-Aldrich
Congo Red, certified by the BSC, BioXtra
Sigma-Aldrich
Congo Red, Dye content ≥35 %
Sigma-Aldrich
Cellulose, fibers, (medium)
Sigma-Aldrich
Cellulase from Trichoderma sp., BioReagent, suitable for plant cell culture, 3-10 units/mg solid
Supelco
Cellulose, acid washed, from spruce, suitable for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, powder, suitable for column chromatography
Supelco
Cellulose, acid washed, powder, suitable for column chromatography
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Carboxymethylcellulose sodium salt, medium viscosity
Sigma-Aldrich
Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
Cellulase from Trichoderma sp., powder, ≥5,000 units/g solid
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9