Merck
  • Development of HPLC method for the determination of levosulpiride in human plasma.
Development of HPLC method for the determination of levosulpiride in human plasma.
Journal of pharmaceutical and biomedical analysis (2004-06-15)
Su-Eon Jin, Eunmi Ban, Yang-Bae Kim, Chong-Kook Kim
ABSTRACT
A rapid and simple high performance liquid chromatography (HPLC) method was developed and validated for determination of levosulpiride in human plasma. After extraction with ethylacetate/methylene chloride (5:1, v/v), analysis of levosulpiride in plasma samples was carried out using a reverse phase C18 column with fluorescence detector (maximum excitation at 300 nm and maximum emission at 365 nm) for separation and quantification. A mixture of methanol-20 mM phosphate buffer (pH 3.5, 16:84, v/v) was used as a mobile phase. The method was specific and sensitive with a limit of quantification of 5 ng/ml. This HPLC method was validated by examining the precision and accuracy for inter- and intra-day analysis in the concentration range of 5-150 ng/ml. The relative standard deviation (R.S.D.) in inter- and intra-day validation were 8.16-19.75 and 3.90-11.69%, respectively. In stability tests, levosulpiride in human plasma was stable during the storage and assay procedure. The method was applied to the bioequivalence study of two levosulpiride tablet formulations (25 mg) after a single oral administration.
Sigma-Aldrich
Butyric acid, ≥99%
Supelco
Butyric acid, analytical standard
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG