Merck
  • Effects of co-exposure to extremely low frequency (ELF) magnetic fields and benzene or benzene metabolites determined in vitro by the alkaline comet assay.
Effects of co-exposure to extremely low frequency (ELF) magnetic fields and benzene or benzene metabolites determined in vitro by the alkaline comet assay.
Toxicology letters (2005-04-20)
Massimo Moretti, Milena Villarini, Stefano Simonucci, Cristina Fatigoni, Giuseppina Scassellati-Sforzolini, Silvano Monarca, Rossana Pasquini, Monica Angelucci, Maila Strappini
ABSTRACT
In the present study, we investigated in vitro the possible genotoxic and/or co-genotoxic activity of 50 Hz (power frequency) magnetic fields (MF) by using the alkaline single-cell microgel-electrophoresis (comet) assay. Sets of experiments were performed to evaluate the possible interaction between 50 Hz MF and the known leukemogen benzene. Three benzene hydroxylated metabolites were also evaluated: 1,2-benzenediol (1,2-BD, catechol), 1,4-benzenediol (1,4-BD, hydroquinone), and 1,2,4-benzenetriol (1,2,4-BT). MF (1 mT) were generated by a system consisting of a pair of parallel coils in a Helmholtz configuration. To evaluate the genotoxic potential of 50 Hz MF, Jurkat cell cultures were exposed to 1 mT MF or sham-exposed for 1h. To evaluate the co-genotoxic activity of MF, the xenobiotics (benzene, catechol, hydroquinone, and 1,2,4-benzenetriol) were added to Jurkat cells subcultures at the beginning of the exposure time. In cell cultures co-exposed to 1 mT (50 Hz) MF, benzene and catechol did not show any genotoxic activity. However, co-exposure of cell cultures to 1 mT MF and hydroquinone led to the appearance of a clear genotoxic effect. Moreover, co-exposure of cell cultures to 1 mT MF and 1,2,4-benzenetriol led to a marked increase in the genotoxicity of the ultimate metabolite of benzene. The possibility that 50 Hz (power frequency) MF might interfere with the genotoxic activity of xenobiotics has important implications, since human populations are likely to be exposed to a variety of genotoxic agents concomitantly with exposure to this type of physical agent.
Sigma-Aldrich
1,2,4-Benzenetriol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Pyrocatechol, ≥99%