Merck
Cell nucleus directed 2,3,5-triiodobenzoic acid conjugates.
Medicinal chemistry (Shariqah (United Arab Emirates)) (2009-08-20)
Alexander Sturzu, Ulrich Vogel, Alireza Gharabaghi, Alexander Beck, Hubert Kalbacher, Hartmut Echner, Stefan Heckl
ABSTRACT
Triiodobenzoic acid (TIBA) represents the core structure of most clinically used contrast agents for computed tomography and other X-ray procedures. To construct an intracellular radiopaque contrast agent, TIBA was coupled to various different positively and negatively charged fluorescein iothiocyanate (FITC)-labelled peptides. TIBA coupled to the SV40 T Antigen nuclear localization sequence (NLS) stained 80% of human glioma cells and caused cell death. This occurred with C- or N-terminal binding of TIBA and with the correct or mutant NLS. No cell death and only small numbers of stained cells (below 3 %) were observed after incubation with NLS conjugates lacking TIBA or after incubation with TIBA-conjugates containing a negatively charged polyglutamic acid stretch. TIBA-conjugates containing the Antennapedia-derived cell-penetrating peptide penetratin were only nuclearly taken up when TIBA and FITC were coupled to lysines outside the 16-amino acid peptide sequence. The study shows that intracellular TIBA may have potential as a chemotherapeutic agent rather than a contrast agent.
Sigma-Aldrich
2,3,5-Triiodobenzoic acid, 98%
Sigma-Aldrich
2,3,5-Triiodobenzoic acid, BioReagent, suitable for plant cell culture, ≥97% (HPLC)