Merck
  • Ligand exchange dynamics in aqueous solution studied with 2DIR spectroscopy.
Ligand exchange dynamics in aqueous solution studied with 2DIR spectroscopy.
The journal of physical chemistry. B (2010-04-30)
Sungnam Park, Minbiao Ji, Kelly J Gaffney
ABSTRACT
We have used time-resolved multidimensional vibrational spectroscopy, generally termed 2DIR spectroscopy, to study the equilibrium dynamics of ligand exchange in an aqueous solution containing 3.4 M Mg(ClO(4))(2) and 1.2 M NaSCN. The sensitivity of the CN stretching frequency of thiocyanate (SCN(-)) to contact ion pair formation with Mg(2+) ions generates distinct spectroscopic signatures for the MgNCS(+) contact ion pair and the free SCN(-). We have utilized 2DIR spectroscopy to successfully resolve the interconversion between these thiocyanate configurations and measured the MgNCS(+) contact ion pair dissociation time constant to be 52 +/- 10 ps. We attribute the observed dynamics to perchlorate-thiocyanate anion exchange in the first solvation shell of the Mg(2+) cation. Magnesium ions in this concentrated ionic solution will be coordinated by water molecules, as well as perchlorate and thiocyanate ions. While prior studies have observed microsecond residence times for water ligands in the first coordination sphere of Mg(2+), our study represents the first experimental observation of anion exchange in the first solvent shell of the Mg(2+) cation. We have also used orientational relaxation and spectral diffusion dynamics to quantify the dynamical distinctions between the free anion and the anion in the contact ion pair.
Sigma-Aldrich
Sodium thiocyanate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium thiocyanate solution, BioUltra, 8 M in H2O
Sigma-Aldrich
Magnesium perchlorate, ACS reagent
Sigma-Aldrich
Magnesium perchlorate, puriss., free-flowing powder, ≥99.0% (calc. based on dry substance, KT)
Sigma-Aldrich
Magnesium perchlorate, puriss. p.a., drying agent, ACS reagent, ≥98.0% (calc. based on dry substance, KT)
Sigma-Aldrich
Sodium thiocyanate, reagent grade, 98-102% (titration)
Sigma-Aldrich
Sodium thiocyanate, ACS reagent, ≥98.0%