Merck
  • Identification of rat cytochromes P450 metabolizing N-(2-methoxyphenyl)hydroxylamine, a human metabolite of the environmental pollutants and carcinogens o-anisidine and o-nitroanisole.
Identification of rat cytochromes P450 metabolizing N-(2-methoxyphenyl)hydroxylamine, a human metabolite of the environmental pollutants and carcinogens o-anisidine and o-nitroanisole.
Neuro endocrinology letters (2010-12-29)
Karel Naiman, Eva Frei, Marie Stiborova
ABSTRACT
N-(2-methoxyphenyl)hydroxylamine is a human metabolite of two industrial and environmental pollutants and bladder carcinogens 2-methoxyaniline (o-anisidine) and 2-methoxynitrobenzene (o-nitroanisole). Metabolism of N-(2-methoxyphenyl)hydroxylamine by rat hepatic microsomes and identification of the major microsomal enzymes participating in this process are aims of this study. HPLC with UV detection was employed for the separation of N-(2-methoxyphenyl)hydroxylamine metabolites. Inducers and inhibitors of microsomal enzymes and rat recombinant CYPs were used to characterize the enzymes participating in N-(2-methoxyphenyl)hydroxylamine metabolism. N-(2-methoxyphenyl)hydroxylamine is metabolized by rat hepatic microsomes predominantly to o-anisidine, the parent carcinogen from which N-(2-methoxyphenyl)hydroxylamine is formed, while o-aminophenol and two N-(2-methoxyphenyl)hydroxylamine metabolites, whose exact structures have not been identified as yet, are minor products. Selective inhibitors of microsomal CYPs, NADPH:CYP reductase and NADH:cytochrome b5 reductase and hepatic microsomes of rats pre-treated with specific inducers of CYPs and NADPH:CYP reductase were used to characterize rat liver microsomal enzymes reducing N-(2-methoxyphenyl)hydroxylamine to o-anisidine. Based on these studies, we attribute most of N-(2-methoxyphenyl)hydroxylamine metabolism to o-anisidine in rat liver to CYP2C, followed by CYP2E1, 2D and 2A. Among recombinant rat CYP enzymes tested in this study, rat CYP2C11 and 2E1, followed by CYP2A2, 2D1/2, 2C12, 3A1/2 and 1A1/2 were the most efficient enzymes metabolizing N-(2-methoxyphenyl)hydroxylamine to o-anisidine. The results found in this study, the first report on the reduction of N-(2-methoxyphenyl)hydroxylamine by rat CYP enzymes, demonstrate that CYP2C, followed by CYP2E1, 2D and 2A are the major enzymes participating in this process in rat liver.
Sigma-Aldrich
2-Nitroanisole, ≥99%
Sigma-Aldrich
o-Anisidine, ≥99%