Merck
Thiourea isosteres as anion receptors and transmembrane transporters.
Chemical communications (Cambridge, England) (2011-06-10)
Marco Wenzel, Mark E Light, Anthony P Davis, Philip A Gale
ABSTRACT
Compounds containing cyanoguanidine and 3-amino-1,2,4-benzothiadiazine-1,1-dioxide have been studied as anion receptors and transporters. Significant affinity for oxo-anions was observed in organic solution and the receptors were found to function as transmembrane chloride/nitrate antiporters with transport rates enhanced in the presence of valinomycin-K(+) complex.
Sigma-Aldrich
Dicyandiamide, 99%
Sigma-Aldrich
Valinomycin, ≥99.0% (TLC)
Supelco
Potassium ionophore I, Selectophore, function tested
Sigma-Aldrich
Valinomycin, ≥98% (TLC), ≥90% (HPLC)
Sigma-Aldrich
Valinomycin, Ready Made Solution, ~1 mg/mL in DMSO