Merck
  • Hydrolysis of N-alkyl sulfamates and the catalytic efficiency of an S-N cleaving sulfamidase.
Hydrolysis of N-alkyl sulfamates and the catalytic efficiency of an S-N cleaving sulfamidase.
The Journal of organic chemistry (2012-02-23)
Danielle C Lohman, Richard Wolfenden, David R Edwards
ABSTRACT
The final step in the degradation of heparin sulfate involves the enzymatic hydrolysis of its 2-sulfamido groups. To evaluate the power of the corresponding sulfamidases as catalysts, we examined the reaction of N-neopentyl sulfamate at elevated temperatures and found it to undergo specific acid catalyzed hydrolysis even at alkaline pH. A rate constant of 10(-16) s(-1) was calculated using the Eyring equation for water attack on the N-protonated species at pH 7, 25 °C. As a model for the pH neutral reaction, a rate constant for hydroxide attack on (CH(3))(3)CCH(2)N(+)H(2)SO(3)(-) at pH 7, 25 °C was calculated to be 10(-19) s(-1). The corresponding rate enhancement (k(cat)/k(non)) produced by the N-sulfamidase of F. heparinum is approximately 10(16)-fold, which is somewhat larger than those generated by most hydrolytic enzymes but considerably smaller than those generated by S-O cleaving sulfatases.
Sigma-Aldrich
Sulfamic acid, 99.999% trace metals basis
Supelco
Sulfamic acid, analytical standard (for acidimetry), ACS reagent
Sigma-Aldrich
Sulfamic acid, ≥99.5% (alkalimetric)
Sigma-Aldrich
Ammonium sulfamate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium sulfamate, ACS reagent, ≥98.0%
Sigma-Aldrich
Sulfamic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Sulfamic acid, ACS reagent, 99.3%
Sigma-Aldrich
Sulfamic acid, reagent grade, 98%