Merck
  • F420H2-dependent degradation of aflatoxin and other furanocoumarins is widespread throughout the actinomycetales.
F420H2-dependent degradation of aflatoxin and other furanocoumarins is widespread throughout the actinomycetales.
PloS one (2012-03-03)
Gauri V Lapalikar, Matthew C Taylor, Andrew C Warden, Colin Scott, Robyn J Russell, John G Oakeshott
ABSTRACT
Two classes of F(420)-dependent reductases (FDR-A and FDR-B) that can reduce aflatoxins and thereby degrade them have previously been isolated from Mycobacterium smegmatis. One class, the FDR-A enzymes, has up to 100 times more activity than the other. F(420) is a cofactor with a low reduction potential that is largely confined to the Actinomycetales and some Archaea and Proteobacteria. We have heterologously expressed ten FDR-A enzymes from diverse Actinomycetales, finding that nine can also use F(420)H(2) to reduce aflatoxin. Thus FDR-As may be responsible for the previously observed degradation of aflatoxin in other Actinomycetales. The one FDR-A enzyme that we found not to reduce aflatoxin belonged to a distinct clade (herein denoted FDR-AA), and our subsequent expression and analysis of seven other FDR-AAs from M. smegmatis found that none could reduce aflatoxin. Certain FDR-A and FDR-B enzymes that could reduce aflatoxin also showed activity with coumarin and three furanocoumarins (angelicin, 8-methoxysporalen and imperatorin), but none of the FDR-AAs tested showed any of these activities. The shared feature of the compounds that were substrates was an α,β-unsaturated lactone moiety. This moiety occurs in a wide variety of otherwise recalcitrant xenobiotics and antibiotics, so the FDR-As and FDR-Bs may have evolved to harness the reducing power of F(420) to metabolise such compounds. Mass spectrometry on the products of the FDR-catalyzed reduction of coumarin and the other furanocoumarins shows their spontaneous hydrolysis to multiple products.
Sigma-Aldrich
Angelicin
Sigma-Aldrich
Imperatorin, ≥98% (HPLC), powder
Supelco
8-Methoxypsoralen, analytical standard
Sigma-Aldrich
Khellin, suitable for microscopy
Supelco
Xanthotoxin, analytical standard