Merck
  • Transforming frozen self-assemblies of amphiphilic block copolymers into dynamic pH-sensitive micelles.
Transforming frozen self-assemblies of amphiphilic block copolymers into dynamic pH-sensitive micelles.
Macromolecular rapid communications (2012-04-26)
Fabien Dutertre, Olivier Boyron, Bernadette Charleux, Christophe Chassenieux, Olivier Colombani
ABSTRACT
The self-assembly in water of an amphiphilic P(nBMA(50%) -stat-DMAEMA(50%) )(100)-b-PDMAEMA(235) diblock copolymer based on hydrophilic dimethylaminoethylmethacrylate (DMAEMA) units and hydrophobic n-butylmethacrylate (nBMA) ones is reported. DMAEMA units have been incorporated into the hydrophobic block of this copolymer to moderate its hydrophobic character. Light scattering experiments revealed the formation of micelles whose apparent aggregation number varied reversibly with the ionization degree of the DMAEMA units. Incorporating hydrophilic units into the hydrophobic block of an amphiphilic block copolymer is thus a way to generate dynamic aggregates in aqueous medium. As this strategy was also successful using other types of hydrophilic units, we believe it to be universal.
Sigma-Aldrich
Anisole, ≥99%, FCC, FG
Sigma-Aldrich
Anisole, anhydrous, 99.7%
Sigma-Aldrich
Anisole, ReagentPlus®, 99%
Supelco
Anisole, analytical standard
Supelco
Anisole, Pharmaceutical Secondary Standard; Certified Reference Material