Merck
  • Nuclear factor kappa B: a pro-inflammatory, transcription factor-mediated signalling pathway in lung carcinogenesis and its inhibition by nonsteroidal anti-inflammatory drugs.
Nuclear factor kappa B: a pro-inflammatory, transcription factor-mediated signalling pathway in lung carcinogenesis and its inhibition by nonsteroidal anti-inflammatory drugs.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer (2012-05-18)
Shruti Setia, Sankar Nath Sanyal
ABSTRACT
9,10-Dimethyl benz(a)anthracene (DMBA), when injected intratracheally once at a dose of 20 mg/kg body weight, is found to induce lung cancer in rats. Two nonsteroidal anti-inflammatory drugs (NSAIDs), indomethacin and etoricoxib, are given orally daily as chemopreventive agents at a dose of 0.6 mg/kg body weight and 2 mg/kg body weight, respectively, along with DMBA. Morphologic and histologic analysis revealed the occurence of tumors and intense cellular proliferation in the DMBA-treated animals, whereas no such features were observed in the other groups. Nuclear factor κB, a nuclear transcription factor, and proliferating cell nuclear antigen, a cell proliferation antigen, were studied by immunoblotting and immunohistochemistry and their levels were markedly elevated in the DMBA group compared with the others. Oxidative stress parameters, as studied by the inducible nitric oxide synthase activity, and the levels of reactive oxygen and nitrogen species were found to be suppressed in the DMBA group. Furthermore, fluorescent staining of the isolated lung cells from bronchoalveolar lavage was performed to study apoptosis and alterations in the mitochondrial membrane potential, and the DMBA-induced lung cancer was found to be associated with high inner mitochondrial membrane potential and a suppressed level of apoptosis.