Merck
  • The role of diallyl sulfides and dipropyl sulfides in the in vitro antimicrobial activity of the essential oil of garlic, Allium sativum L., and leek, Allium porrum L.
The role of diallyl sulfides and dipropyl sulfides in the in vitro antimicrobial activity of the essential oil of garlic, Allium sativum L., and leek, Allium porrum L.
Phytotherapy research : PTR (2012-05-23)
Sergio Casella, Michele Leonardi, Bernardo Melai, Filippo Fratini, Luisa Pistelli
ABSTRACT
The in vitro antibacterial activity of essential oils (EOs) obtained from fresh bulbs of garlic, Allium sativum L., and leek, Allium porrum L. ( Alliaceae), was studied. A. sativum (garlic) EO showed a good antimicrobial activity against Staphylococcus aureus (inhibition zone 14.8 mm), Pseudomonas aeruginosa (inhibition zone 21.1 mm), and Escherichia coli (inhibition zone 11.0 mm), whereas the EO of A. porrum (leek) had no antimicrobial activity. The main constituents of the garlic EO were diallyl monosulfide, diallyl disulfide (DADS), diallyl trisulfide, and diallyl tetrasulfide. The EO of A. porrum was characterized by the presence of dipropyl disulfide (DPDS), dipropyl trisulfide, and dipropyl tetrasulfide. The antimicrobial activities of the DADS and DPDS were also studied. The results obtained suggest that the presence of the allyl group is fundamental for the antimicrobial activity of these sulfide derivatives when they are present in Allium or in other species (DADS inhibition zone on S. aureus 15.9 mm, P. aeruginosa 21.9 mm, E. coli 11.4 mm).
Sigma-Aldrich
Allyl sulfide, ≥97%, FG
Sigma-Aldrich
Allyl sulfide, 97%
Sigma-Aldrich
Allyl disulfide, ≥80%, FG
Sigma-Aldrich
Allyl disulfide, technical grade, 80%