Merck
  • Patch testing with the textile dyes Disperse Orange 1 and Disperse Yellow 3 and some of their potential metabolites, and simultaneous reactions to para-amino compounds.
Patch testing with the textile dyes Disperse Orange 1 and Disperse Yellow 3 and some of their potential metabolites, and simultaneous reactions to para-amino compounds.
Contact dermatitis (2012-05-26)
Laura Malinauskiene, Erik Zimerson, Magnus Bruze, Kristina Ryberg, Marlene Isaksson
ABSTRACT
It is known that, in vitro, human skin bacteria are able to split disperse azo dyes into the corresponding aromatic amines, some of which are sensitizers in the local lymph node assay. We hypothesize that the molecules of disperse dyes migrate onto the skin while garments are worn, and are metabolized and degraded by commensal skin bacteria. These molecules penetrate the skin and induce sensitization. To evaluate the elicitation capacities of the possible azo-degradation products of the selected azo disperse dyes in patients allergic to them and to compare it with the elicitation capacities of other para-compounds. Ten patients allergic to Disperse Yellow 3 (DY3) and/or Disperse Orange 1 (DO1) were patch tested with a dilution series of the purified dyes 4-nitroaniline and p-aminodiphenylamine in concentrations equimolar to those of purified DO1 in the dilution series, as well as 4-aminoacetanilide and 2-amino-p-cresol in concentrations equimolar to those of purified DY3 in the dilution series. Results. Three patterns of patch test reactions could be seen. The 6 patients who were positive to DO1 and DY3 also reacted to p-aminodiphenylamine and 2-amino-p-cresol. Two patients were positive to DO1 only, and both reacted to p-aminodiphenylamine, but to neither 4-aminoacetanilide or 2-amino-p-cresol. Two patients did not react to DO1 or DY3 on this occasion. We show that it is possible that the major sensitizers in contact allergy to DO1 and DY3 are their metabolites, p-aminodiphenylamine and 2-amino-p-cresol, respectively, which might be formed by the azoreductase pathway of skin bacteria.
Sigma-Aldrich
p-Phenylenediamine, sublimed, ≥99%
Supelco
Disperse Orange 1, analytical standard
Supelco
Disperse Yellow 3, analytical standard
Sigma-Aldrich
Disperse Yellow 3, Dye content 30 %
Sigma-Aldrich
p-Phenylenediamine, ≥99.0% (GC/NT)
Sigma-Aldrich
p-Phenylenediamine, 98% (GC)
Sigma-Aldrich
2-Amino-4-methylphenol, 97%
Supelco
4-Nitroaniline, analytical standard
Sigma-Aldrich
4-Nitroaniline, ≥99%
Sigma-Aldrich
1,4-Phenylenediamine dihydrochloride, ≥99.0%