Merck
  • The TetR-type transcriptional repressor RolR from Corynebacterium glutamicum regulates resorcinol catabolism by binding to a unique operator, rolO.
The TetR-type transcriptional repressor RolR from Corynebacterium glutamicum regulates resorcinol catabolism by binding to a unique operator, rolO.
Applied and environmental microbiology (2012-06-19)
Tang Li, Kexin Zhao, Yan Huang, Defeng Li, Cheng-Ying Jiang, Nan Zhou, Zheng Fan, Shuang-Jiang Liu
ABSTRACT
The rol (designated for resorcinol) gene cluster rolRHMD is involved in resorcinol catabolism in Corynebacterium glutamicum, and RolR is the TetR-type regulator. In this study, we investigated how RolR regulated the transcription of the rol genes in C. glutamicum. The transcription start sites and promoters of rolR and rolHMD were identified. Quantitative reverse transcription-PCR and promoter activity analysis indicated that RolR negatively regulated the transcription of rolHMD and of its own gene. Further, a 29-bp operator rolO was located at the intergenic region of rolR and rolHMD and was identified as the sole binding site for RolR. It contained two overlapping inverted repeats and they were essential for RolR-binding. The binding of RolR to rolO was affected by resorcinol and hydroxyquinol, which are the starting compounds of resorcinol catabolic pathway. These two compounds were able to dissociate RolR-rolO complex, thus releasing RolR from the complex and derepressing the transcription of rol genes in C. glutamicum. It is proposed that the binding of RolR to its operator rolO blocks the transcription of rolHMD and of its own gene, thus negatively regulated resorcinol degradation in C. glutamicum.
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
Supelco
Resorcinol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
1,2,4-Benzenetriol, ReagentPlus®, 99%