Merck
  • Self-assembled Pd6 open cage with triimidazole walls and the use of its confined nanospace for catalytic Knoevenagel- and Diels-Alder reactions in aqueous medium.
Self-assembled Pd6 open cage with triimidazole walls and the use of its confined nanospace for catalytic Knoevenagel- and Diels-Alder reactions in aqueous medium.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-08-18)
Dipak Samanta, Sandip Mukherjee, Yogesh P Patil, Partha Sarathi Mukherjee
ABSTRACT
The two-component self-assembly of a 90° Pd(II) acceptor and a triimidazole donor led to the formation of a water-soluble semi-cylindrical cage with a hydrophobic cavity, which was separately crystallized with hydrophilic- and hydrophobic guests. The parent cage was found to catalyze the Knoevenagel condensation reaction of a series of aromatic mono-aldehydes with active methylene compounds, such as Meldrum's acid or 1,3-dimethylbarbituric acid. The confined hydrophobic nanospace within this cage was also used in the catalytic Diels-Alder reactions of 9-hydroxymethylanthracene with N-phenylmaleimide or N-cyclohexylmaleimide.
Sigma-Aldrich
9-Anthracenemethanol, 97%
Sigma-Aldrich
N-Cyclohexylmaleimide, 97%
Sigma-Aldrich
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%