Merck
  • Gymnothelignans A-O: conformation and absolute configuration analyses of lignans bearing tetrahydrofuran from Gymnotheca chinensis.
Gymnothelignans A-O: conformation and absolute configuration analyses of lignans bearing tetrahydrofuran from Gymnotheca chinensis.
The Journal of organic chemistry (2012-09-06)
Dahai He, Lisheng Ding, Hongxi Xu, Xinxiang Lei, Hongping Xiao, Yan Zhou
ABSTRACT
Fifteen new lignans, gymnothelignans A-O (1-15), bearing tetrahydrofuran with variable conformations belonging to three potentially related skeletons were isolated from Gymnotheca chinensis Decne. The structures were elucidated by means of detailed spectroscopic analysis. Absolute configurations were assigned using X-ray single-crystal diffraction and chemical transformations. Moreover, by the homology, compounds 1-11 and eupomatilones were confirmed to have uniform R-configuration at C-5. However, a synthesized congener has long been mistaken as 5-epimer of eupomatilone-6. This work provides guidance for the absolute configuration establishment of the subeupomatilone family with trans-H-4-H-5 configuration.
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Supelco
Tetrahydrofuran, analytical standard