Merck
  • Influence of the pore structure and surface chemistry on adsorption of ethylbenzene and xylene isomers by KOH-activated multi-walled carbon nanotubes.
Influence of the pore structure and surface chemistry on adsorption of ethylbenzene and xylene isomers by KOH-activated multi-walled carbon nanotubes.
Journal of hazardous materials (2012-09-12)
Fei Yu, Yanqing Wu, Jie Ma
ABSTRACT
To investigate the effect of pore structure and surface chemistry on adsorption of monoaromatics, multi-walled carbon nanotubes (MWCNTs) with different specific surface area (SSA) and surface oxygen content by KOH activation treatment have been prepared. The results indicate that adsorption of four test chemicals is enhanced by 2-3, 7-9 and 10-13 times for CNTs-K1 (MWCNTs:KOH=1:1), CNTs-K4 (MWCNTs:KOH=1:4) and CNTs-K6 (MWCNTs:KOH=1:6) as justified by the ratios of adsorption coefficient. Adsorption properties of ethylbenzene and xylene isomers (EX) by activated-MWCNTs depend significantly on mesopore volumes and hydroxyl groups. There are strong positive correlations between the adsorption capacity (or adsorption affinity) and mesopore volumes or hydroxyl groups. Mesopore-filling could be a predominant mechanism governing monoarmatics adsorption onto activated-MWCNTs, contributing more than simply increasing SSA. The hydroxyl groups at low concentration can improve diffusion of MWCNTs in water, which could be favorable for aqueous phase adsorption. However, it is not large enough to form a lot of water clusters to hinder EX adsorption. This is the first systematic study of the influence of physicochemical properties on adsorption of monoaromatics onto activated-MWCNTs by qualitative and quantitative analysis, and results highlight the synergetic effects of porosity and surface chemistry in controlling adsorption of monoaromatics onto MWCNTs in wastewater treatment.
Product Description
Supelco
Ethylbenzene solution, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 5% in chloroform-d (99.8 atom % D), TMS 1 %, NMR tube size 5 mm × 7 in.
Supelco
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 8 mm × 8 in.
Supelco
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d ("100%", 99.96 atom % D)
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 1% in chloroform-d (99.8 atom % D)
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%
Supelco
Ethylbenzene, analytical standard
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Ethylbenzene, ReagentPlus®, 99%
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets