Merck
  • Silica gel-mediated organic reactions under organic solvent-free conditions.
Silica gel-mediated organic reactions under organic solvent-free conditions.
Molecules (Basel, Switzerland) (2012-09-29)
Satoaki Onitsuka, Yong Zhi Jin, Ajam C Shaikh, Hiroshi Furuno, Junji Inanaga
ABSTRACT
Silica gel was found to be an excellent medium for some useful organic transformations under organic solvent-free conditions, such as (1) the Friedel-Crafts-type nitration of arenes using commercial aqueous 69% nitric acid alone at room temperature, (2) one-pot Wittig-type olefination of aldehydes with activated organic halides in the presence of tributyl- or triphenylphosphine and Hunig's base, and (3) the Morita-Baylis-Hillman reaction of aldehydes with methyl acrylate. After the reactions, the desired products were easily obtained in good to excellent yields through simple manipulation.
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥65%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%