Merck
  • Age-dependent effects of systemic administration of oxytetracycline on the viscoelastic properties of rat tail tendons as a mechanistic basis for pharmacological treatment of flexural limb deformities in foals.
Age-dependent effects of systemic administration of oxytetracycline on the viscoelastic properties of rat tail tendons as a mechanistic basis for pharmacological treatment of flexural limb deformities in foals.
American journal of veterinary research (2012-11-28)
Leslie R Wintz, Michael Lavagnino, Keri L Gardner, Aleksa M Sedlak, Steven P Arnoczky
ABSTRACT
To describe the effect of systemically administered oxytetracycline on the viscoelastic properties of rat tail tendon fascicles (TTfs) to provide a mechanistic rationale for pharmacological treatment of flexural limb deformities in foals. TTfs from ten 1-month-old and ten 6-month-old male Sprague-Dawley rats. 5 rats in each age group were administered oxytetracycline (50 mg/kg, IP, q 24 h) for 4 days. The remaining 5 rats in each age group served as untreated controls. Five days after initiation of oxytetracycline treatment, TTfs were collected and their viscoelastic properties were evaluated via a stress-relaxation protocol. Maximum modulus and equilibrium modulus were compared via a 2-way ANOVA. Collagen fibril size, density, and orientation in TTfs were compared between treated and control rats. Viscoelastic properties were significantly decreased in TTfs from 1-month-old oxytetracycline-treated rats, compared with those in TTfs from 1-month-old control rats. Oxytetracycline had no effect on the viscoelastic properties of TTfs from 6-month-old rats. Collagen fibril size, density, and orientation in TTfs from 1-month-old rats did not differ between oxytetracycline-treated and control rats. Results confirmed that systemically administered oxytetracycline decreased the viscoelastic properties of TTfs from 1-month-old rats but not those of TTfs from 6-month-old rats. The decrease in viscoelastic properties associated with oxytetracycline treatment does not appear to be caused by altered collagen fibril diameter or organization. The age-dependent effect of oxytetracycline on the viscoelastic properties of tendons may be related to its effect on the maturation of the extracellular matrix of developing tendons.
Product Description
Millipore
Oxytetra Selective Supplement, suitable for microbiology
Sigma-Aldrich
Oxytetracycline hydrochloride
Sigma-Aldrich
Oxytetracycline dihydrate, Assay (anhydrous basis) 95.0%-102.0%
Supelco
Oxytetracycline hydrochloride, VETRANAL®, analytical standard
Oxytetracycline hydrochloride, European Pharmacopoeia (EP) Reference Standard