Merck
Biovanillin from agro wastes as an alternative food flavour.
Journal of the science of food and agriculture (2012-12-05)
Nur Ain Zamzuri, Suraini Abd-Aziz
ABSTRACT
This review provides an overview of biovanillin production from agro wastes as an alternative food flavour. Biovanillin is one of the widely used flavour compounds in the foods, beverages and pharmaceutical industries. An alternative production approach for biovanillin as a food flavour is hoped for due to the high and variable cost of natural vanillin as well as the limited availability of vanilla pods in the market. Natural vanillin refers to the main organic compound that is extracted from the vanilla bean, as compared to biovanillin, which is produced biologically by microorganisms from a natural precursor such as ferulic acid. Biovanillin is also reviewed as a potential bioflavour produced by microbial fermentation in an economically feasible way in the near future. In fact, we briefly discuss natural, synthetic and biovanillin and the types of agro wastes that are useful as sources for bioconversion of ferulic acid into biovanillin. The subsequent part of the review emphasizes the current application of vanillin as well as the utilization of biovanillin as an alternative food flavour. The final part summarizes biovanillin production from agro wastes that could be of benefit as a food flavour derived from potential natural precursors.
Product Description
Supelco
Isoeugenol, analytical standard
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Isoeugenol, 98%, mixture of cis and trans
Sigma-Aldrich
Isoeugenol, mixture of cis and trans, 99%, FG
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Vanillin, European Pharmacopoeia (EP) Reference Standard
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Supelco
trans-Ferulic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland