Merck
  • Enantiomerically pure and racemic dimethyl{N-[(2-oxidonaphthalen-1-yl-κO)methylidene]valinato-κ2N,O}tin(IV).
Enantiomerically pure and racemic dimethyl{N-[(2-oxidonaphthalen-1-yl-κO)methylidene]valinato-κ2N,O}tin(IV).
Acta crystallographica. Section C, Crystal structure communications (2013-01-04)
Uwe Böhme, Sabine Fels
ABSTRACT
The title compound, [Sn(CH(3))(2)(C(16)H(15)NO(3))], crystallized from one reaction batch with high enantiomeric excess as both a pure enantiomer and a racemate. The S enantiomer crystallizes in the chiral space group P3(2). The racemate crystallizes in the space group P1 with R and S enantiomers in the crystal lattice; these form dimers about a crystallographic inversion centre.
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, ≥99%, powder