Merck
  • A divergent approach to the synthesis of the yohimbinoid alkaloids venenatine and alstovenine.
A divergent approach to the synthesis of the yohimbinoid alkaloids venenatine and alstovenine.
Nature chemistry (2013-01-25)
Terry P Lebold, Jessica L Wood, Josh Deitch, Michael W Lodewyk, Dean J Tantillo, Richmond Sarpong
ABSTRACT
The yohimbinoid alkaloids continue to receive considerable attention from the synthetic community because of their interesting chemical structures and varied biological activity. Although there are several elegant syntheses of certain members of this group of alkaloids, a truly unified approach has yet to be developed. In short, general approaches to this compound class are hampered by a lack of complete control in setting the C(3) stereocentre at a late stage. Herein, we report that a functionalized hydrindanone enables a divergent strategy that builds on existing precedent to address this long-standing challenge. Utilizing an aminonitrile intermediate, the stereochemistry at C(3) of the yohimbinoid skeleton can be controlled effectively in a Pictet-Spengler reaction. We applied this approach to the first total syntheses of the C(3) epimeric natural products venenatine and alstovenine.
Product Description
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%