Merck
  • Ultralow-limit gas detection in nano-dumbbell polymer sensor via electrospinning.
Ultralow-limit gas detection in nano-dumbbell polymer sensor via electrospinning.
Nanoscale (2013-02-02)
Mianqi Xue, Fengwang Li, Yue Wang, Xiaojing Cai, Feng Pan, Jitao Chen
ABSTRACT
Nano-dumbbells via electrospinning: controllable nano-dumbbells are fabricated via electrospinning. The weight block parts and the length of nanowires between them can be adjusted through changing the experimental conditions. This nanostructure enables ultralow-limit gas sensing properties of the resulting polypyrrole-based microsensor.
Sigma-Aldrich
Kollidon® 25
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Povidone, meets USP testing specifications
Povidone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Polypyrrole, doped, conductivity 0.5-1.5 S/cm (pressed pellet, typical), extent of labeling: ~5 wt. % loading, coated on titanium dioxide
Sigma-Aldrich
Polypyrrole, conductivity 10-50 S/cm (pressed pellet)
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, Molecular Biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polypyrrole, doped, conductivity 30 S/cm (bulk), extent of labeling: 20 wt. % loading, composite with carbon black
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000