Merck
  • Non-adiabatic ab initio molecular dynamics of supersonic beam epitaxy of silicon carbide at room temperature.
Non-adiabatic ab initio molecular dynamics of supersonic beam epitaxy of silicon carbide at room temperature.
The Journal of chemical physics (2013-02-08)
Simone Taioli, Giovanni Garberoglio, Stefano Simonucci, Silvio a Beccara, Lucrezia Aversa, Marco Nardi, Roberto Verucchi, Salvatore Iannotta, Maurizio Dapor, Dario Alfè
ABSTRACT
In this work, we investigate the processes leading to the room-temperature growth of silicon carbide thin films by supersonic molecular beam epitaxy technique. We present experimental data showing that the collision of fullerene on a silicon surface induces strong chemical-physical perturbations and, for sufficient velocity, disruption of molecular bonds, and cage breaking with formation of nanostructures with different stoichiometric character. We show that in these out-of-equilibrium conditions, it is necessary to go beyond the standard implementations of density functional theory, as ab initio methods based on the Born-Oppenheimer approximation fail to capture the excited-state dynamics. In particular, we analyse the Si-C(60) collision within the non-adiabatic nuclear dynamics framework, where stochastic hops occur between adiabatic surfaces calculated with time-dependent density functional theory. This theoretical description of the C(60) impact on the Si surface is in good agreement with our experimental findings.
Sigma-Aldrich
Silicon carbide, nanopowder, <100 nm particle size
Sigma-Aldrich
Silicon carbide, −400 mesh particle size, ≥97.5%
Sigma-Aldrich
Silicon carbide, -200 mesh particle size