Merck
  • Effect of titanium preoxidation on wrought pure titanium to ceramic bond strength.
Effect of titanium preoxidation on wrought pure titanium to ceramic bond strength.
The Journal of prosthetic dentistry (2013-02-12)
Cui-cui Zhang, Jian-tao Ye, Yi-ping Zhang, Juan-kun Liao, Bo-hua Li
ABSTRACT
The creation of high bond strength between machined computer-manufactured pure titanium and porcelain remains a problem. However, machined titanium does not form the thick titanium oxide film found in cast titanium. The purpose of this study was to investigate the effects of different preoxidation treatments on the bond strength of a machined pure titanium ceramic system. Specimens of commercially pure titanium (25 × 3 × 0.5 mm) were divided equally into 6 groups (n=8), which received different preoxidation treatments (3 hour natural oxidation; 600°C, 650°C, 700°C, 750°C, and 800°C for 3 minutes). Bond strengths were evaluated by using a 3-point bend test. The results were analyzed by using 1-way ANOVA and the least significant difference test. Twelve additional specimens of commercially pure titanium (15 × 3 × 0.5 mm) were cut for interface observation and divided equally into 6 groups that received the preoxidation treatments described previously. Scanning electron microscopy and energy dispersive spectrum were used to observe microscopic features of the interface between Ti and ceramic. The bond strength values of the 6 groups ranged from 23.72 ±2.53 MPa to 36.99 ±3.92 MPa, with significant differences (P<.05). The specimen that received 750°C preoxidation had the highest bond strength. The main interface elements of the 6 groups were O, Si, Ti, Sn, Al, Na, and K. Ti showed a sigmoidal diffusion curve in each group, and Si showed a sigmoidal diffusion curve in most groups. Sn was enriched in each group's interface. Preoxidation under vacuum before porcelain firing can effectively improve the bond strength of machined pure titanium-porcelain systems.
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis