Merck
  • Kainate receptor post-translational modifications differentially regulate association with 4.1N to control activity-dependent receptor endocytosis.
Kainate receptor post-translational modifications differentially regulate association with 4.1N to control activity-dependent receptor endocytosis.
The Journal of biological chemistry (2013-02-13)
Bryan A Copits, Geoffrey T Swanson
ABSTRACT
Kainate receptors exhibit a highly compartmentalized distribution within the brain; however, the molecular and cellular mechanisms that coordinate their expression at neuronal sites of action are poorly characterized. Here we report that the GluK1 and GluK2 kainate receptor subunits interact with the spectrin-actin binding scaffolding protein 4.1N through a membrane-proximal domain in the C-terminal tail. We found that this interaction is important for the forward trafficking of GluK2a receptors, their distribution in the neuronal plasma membrane, and regulation of receptor endocytosis. The association between GluK2a receptors and 4.1N was regulated by both palmitoylation and protein kinase C (PKC) phosphorylation of the receptor subunit. Palmitoylation of the GluK2a subunit promoted 4.1N association, and palmitoylation-deficient receptors exhibited reduced neuronal surface expression and compromised endocytosis. Conversely, PKC activation decreased 4.1N interaction with GluK2/3-containing kainate receptors in acute brain slices, an effect that was reversed after inhibition of PKC. Our data and previous studies therefore demonstrate that these two post-translational modifications have opposing effects on 4.1N association with GluK2 kainate and GluA1 AMPA receptors. The convergence of the signaling pathways regulating 4.1N protein association could thus result in the selective removal of AMPA receptors from the plasma membrane while simultaneously promoting the insertion and stabilization of kainate receptors, which may be important for tuning neuronal excitability and synaptic plasticity.
Product Description
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Anti-Myc Tag Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-GluR1-NT (NT) Antibody, clone RH95, clone RH95, from mouse
Sigma-Aldrich
Anti-GluR6/7 Antibody, clone NL9, rabbit monoclonal, culture supernatant, clone NL904, Upstate®