Merck
Turn-on, fluorescent nuclear stains with live cell compatibility.
Organic letters (2013-03-07)
Demar R G Pitter, Jens Wigenius, Adrienne S Brown, James D Baker, Fredrik Westerlund, James N Wilson
ABSTRACT
DNA-binding, green and yellow fluorescent probes with excellent brightness and high on/off ratios are reported. The probes are membrane permeable, live-cell compatible, and optimally matched to 405 nm and 514 nm laser lines, making them attractive alternatives to UV-excited and blue emissive Hoechst 33342 and DAPI nuclear stains. Their electronic structure was investigated by optical spectroscopy supported by TD-DFT calculations. DNA binding is accompanied by 27- to 75-fold emission enhancements, and linear dichroism demonstrates that one dye is a groove binder while the other intercalates into DNA.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, suitable for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, suitable for fluorescence, BioReagent, ≥95.0% (HPLC)
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC), suitable for fluorescence