Merck
  • New surface-enhanced Raman scattering platforms: composite calcium carbonate microspheres coated with astralen and silver nanoparticles.
New surface-enhanced Raman scattering platforms: composite calcium carbonate microspheres coated with astralen and silver nanoparticles.
Langmuir : the ACS journal of surfaces and colloids (2013-03-09)
Inna Y Stetciura, Alexey V Markin, Andrei N Ponomarev, Alexander V Yakimansky, Tatiana S Demina, Cristian Grandfils, Dmitry V Volodkin, Dmitry A Gorin
ABSTRACT
Surface-enhanced Raman scattering (SERS) microspectroscopy is a very promising label-free, noncontact, and nondestructive method for real-time monitoring of extracellular matrix (ECM) development and cell integration in scaffolds for tissue engineering. Here, we prepare a new type of micrometer-sized SERS substrate, core-shell microparticles composed of solid carbonate core coated with silver nanoparticles and polyhedral multishell fullerene-like structure, astralen. Astralen has been assembled with polyallylamine hydrochloride (PAH) by the layer-by-layer manner followed by Ag nanoparticle formation by means of a silver mirror reaction, giving the final structure of composite particles CaCO3(PAH/astralen)x/Ag, where x = 1-3. The components of the microparticle carry multiple functionalities: (i) an easy identification by Raman imaging (photostable astralen) and (ii) SERS due to a rough surface of Ag nanoparticles. A combination of Ag and astralen nanoparticles provides an enhancement of astralen Raman signal by more than 1 order of magnitude. Raman signals of commonly used scaffold components such as polylactide and polyvinyl alcohol as well as ECM component (hyaluronic acid) are significantly enhanced. Thus, we demonstrate that new mechanically robust and easily detectable (by astralen signal or optically) core-shell microspheres based on biocompatible CaCO3 can be used as SERS platform. Particle design opens many future perspectives for fabrication of SERS platforms with multiple functions for biomedical applications, for example, for theranostic.
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 8,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 150,000 (average), 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Poly(allylamine hydrochloride), average Mw ~17,500 (GPC vs. PEG std.)
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)