Merck
  • Dimethyl sulfoxide at high concentrations inhibits non-selective cation channels in human erythrocytes.
Dimethyl sulfoxide at high concentrations inhibits non-selective cation channels in human erythrocytes.
General physiology and biophysics (2013-03-28)
Oleg A Nardid, Miroslav I Schetinskey, Yuliya V Kucherenko
ABSTRACT
Dimethyl sulfoxide (DMSO), a by-product of the pulping industry, is widely used in biological research, cryobiology and medicine. On cellular level DMSO was shown to suppress NMDA-AMPA channels activation, blocks Na+ channel activation and attenuates Ca2+ influx (Lu and Mattson 2001). In the present study we explored the whole-cell patch-clamp to examine the acute effect of high concentrations of DMSO (0.1-2 mol/l) on cation channels activity in human erythrocytes. Acute application of DMSO (0.1-2 mol/l) dissolved in Cl--containing saline buffer solution significantly inhibited cation conductance in human erythrocytes. Inhibition was concentration-dependent and had an exponential decay profile. DMSO (2 mol/l) induced cation inhibition in Cl-- containing saline solutions of: 40.3 ± 3.9% for K+, 35.4 ± 3.1% for Ca2+ and 47.4 ± 1.9% for NMDG+. Substitution of Cl- with gluconate- increased the inhibitory effect of DMSO on the Na+ current. Inhibitory effect of DMSO was neither due to high permeability of erythrocytes to DMSO nor to an increased tonicity of the bath media since no effect was observed in 2 mol/l glycerol solution. In conclusion, we have shown that high concentrations of DMSO inhibit the non-selective cation channels in human erythrocytes and thus protect the cells against Na+ and Ca2+ overload. Possible mechanisms of DMSO effect on cation conductance are discussed.
Product Description
Supelco
Dimethyl Sulfoxide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Gluconic acid solution, 49-53 wt. % in H2O
Sigma-Aldrich
Sodium gluconate, meets USP testing specifications
Sigma-Aldrich
D-Gluconic acid sodium salt, ≥99% (HPLC)
Supelco
D-Gluconic acid sodium salt, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Potassium gluconate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Magnesium D-gluconate hydrate, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Potassium gluconate, 97.0-103.0% dry basis, meets USP testing specifications
Sigma-Aldrich
Potassium D-gluconate, ≥99% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Magnesium D-gluconate hydrate, 98.0-102% anhydrous basis, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Iron(II) D-gluconate dihydrate, 98%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%