Merck
Teaching model for artificial teeth and endodontic apex locators.
Journal of dental education (2013-05-10)
Jörg Philipp Tchorz, Elmar Hellwig, Markus Jörg Altenburger
ABSTRACT
Artificial teeth are a useful teaching aid during endodontic education. This article describes the development of a simple and inexpensive model that can be equipped with artificial teeth. It shows that working length determination in artificial root canals using electronic apex locators is possible and that the embedding media has no influence on measurements. The model supports the application of current endodontic techniques and facilitates a validated evaluation of the treatments between students. Artificial teeth can be removed for visualization and replaced for further endodontic exercises.
Sigma-Aldrich
Drierite, with indicator, 4 mesh
Sigma-Aldrich
Calcium sulfate hemihydrate, purum, ≥97.0%
Sigma-Aldrich
Drierite, without indicator, 8 mesh
Sigma-Aldrich
Drierite, without indicator, 4 mesh
Sigma-Aldrich
Drierite, with indicator, 8 mesh
Sigma-Aldrich
Drierite, with indicator, 10-20 mesh
Sigma-Aldrich
Drierite, with indicator, 6 mesh
Sigma-Aldrich
Calcium sulfate dihydrate, puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
Sigma-Aldrich
Calcium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium sulfate dihydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium sulfate, −325 mesh, 99%
Sigma-Aldrich
Calcium sulfate dihydrate, ACS reagent, 98%