Merck
  • [Optimum heart rate - the current goal of cardiovascular therapy. Position statement of the Polish Cardiac Society Working Group on Cardiovascular Drug Therapy].
[Optimum heart rate - the current goal of cardiovascular therapy. Position statement of the Polish Cardiac Society Working Group on Cardiovascular Drug Therapy].
Kardiologia polska (2012-10-20)
Jarosław D Kasprzak, Janina Stępińska, Beata Wożakowska Kapłon, Jarosław Drożdż, Stefan Grajek, Grzegorz Opolski, Andrzej Rynkiewicz, Andrzej Tykarski, Krzysztof J Filipiak
Sigma-Aldrich
(+)-cis-Diltiazem hydrochloride, ≥99% (HPLC)
Diltiazem for system suitability, European Pharmacopoeia (EP) Reference Standard
Diltiazem hydrochloride, European Pharmacopoeia (EP) Reference Standard