Merck
  • Repeated moderate-dose ethanol bouts impair cognitive function in Wistar rats.
Repeated moderate-dose ethanol bouts impair cognitive function in Wistar rats.
Addiction biology (2011-10-27)
Alexander Kuzmin, Sture Liljequist, Jennifer Meis, Vladimir Chefer, Toni Shippenberg, Georgy Bakalkin
ABSTRACT
The effects of repeated, intermittent administration of a moderate dose of ethanol (3.4 g/kg/day × 6 days, intragastrically via gavages) on cognitive function were examined in male Wistar rats. No significant differences in weight gain between the ethanol- and water-treated rats were found. Analysis of physical dependence revealed no signs of spontaneous withdrawal, whereas withdrawal signs exacerbated by Ro15-4513, an inverse benzodiazepine agonist, were apparent 5 hours but not 24 hours after the cessation of ethanol treatment. Spatial learning and memory, as assessed in the Barnes maze, were impaired 3-6 days following the treatment but recovered by the 11th-14th days. Reversal learning, however, was impaired throughout the 2-week observation period. Thus, bouts of moderate-dose ethanol administration transiently impair spatial learning and memory, and promote cognitive inflexibility. The employed ethanol exposure paradigm may provide a model of human cognitive deficits associated with alcohol binge drinking.