Merck
  • EDTA-dependent pseudothrombocytopenia complicated by eosinophilic pneumonia.
EDTA-dependent pseudothrombocytopenia complicated by eosinophilic pneumonia.
The American journal of emergency medicine (2013-05-25)
Naoko Ohashi, Kensuke Nakamura, Ryota Inokuchi, Hajime Sato, Kurato Tokunaga, Tatsuma Fukuda, Susumu Nakajima, Naoki Yahagi
ABSTRACT
EDTA-dependent pseudothrombocytopenia (EDTA-PTCP) is a phenomenon that occurs in vitro when EDTA reacts with harvested blood. EDTA-dependent pseudothrombocytopenia usually does not indicate thrombocytopenia in vivo. Here, we report the first case of EDTA-PTCP complicated by eosinophilic pneumonia. A 70-year-old man with rectal cancer was admitted to the hospital for a liver abscess and rectal cancer. At the time of admission, his platelet count was 20,000/μL, but a peripheral blood smear showed platelet aggregation and the platelet count for a kanamycin-added EDTA blood sample was 180,000/μL. The patient's respiratory status worsened after treatment for the liver abscess and rectal cancer. The patient's bronchoalveolar lavage contained 45% eosinophils, and a diagnosis of acute eosinophilic pneumonia was made. In recent studies, the occurrence of eosinophilic disease has been shown in idiopathic thrombocytopenic purpura. EDTA-dependent pseudothrombocytopenia is an in vitro phenomenon, although platelet activation that results in eosinophil invasion may occur in severe cases.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)