Merck
  • Iron bioavailability from a lipid-based complementary food fortificant mixed with millet porridge can be optimized by adding phytase and ascorbic acid but not by using a mixture of ferrous sulfate and sodium iron EDTA.
Iron bioavailability from a lipid-based complementary food fortificant mixed with millet porridge can be optimized by adding phytase and ascorbic acid but not by using a mixture of ferrous sulfate and sodium iron EDTA.
The Journal of nutrition (2013-06-14)
Colin I Cercamondi, Ines M Egli, Evariste Mitchikpe, Felicien Tossou, Joamel Hessou, Christophe Zeder, Joseph D Hounhouigan, Richard F Hurrell
ABSTRACT
Home fortification with lipid-based nutrient supplements (LNSs) is a promising approach to improve bioavailable iron and energy intake of young children in developing countries. To optimize iron bioavailability from an LNS named complementary food fortificant (CFF), 3 stable isotope studies were conducted in 52 young Beninese children. Test meals consisted of millet porridge mixed with CFF and ascorbic acid (AA). Study 1 compared iron absorption from FeSO4-fortifed meals with meals fortified with a mixture of FeSO4 and NaFeEDTA. Study 2 compared iron absorption from FeSO4-fortifed meals without or with extra AA. Study 3 compared iron absorption from FeSO4-fortified meals with meals containing phytase added prior to consumption, once without or once with extra AA. Iron absorption was measured as erythrocyte incorporation of stable isotopes. In study 1, iron absorption from FeSO4 (8.4%) was higher than that from the mixture of NaFeEDTA and FeSO4 (5.9%; P < 0.05). In study 2, the extra AA increased absorption (11.6%) compared with the standard AA concentration (7.3%; P < 0.001). In study 3, absorption from meals containing phytase without or with extra AA (15.8 and 19.9%, respectively) increased compared with meals without phytase (8.0%; P < 0.001). The addition of extra AA to meals containing phytase increased absorption compared with the test meals containing phytase without extra AA (P < 0.05). These findings suggest that phytase and AA, and especially a combination of the two, but not a mixture of FeSO4 and NaFeEDTA would be useful strategies to increase iron bioavailability from a CFF mixed with cereal porridge.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid iron(III) sodium salt, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Iron(II) sulfate heptahydrate, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid ferric sodium salt, suitable for plant cell culture, BioReagent, crystalline
Sigma-Aldrich
Ferritin from equine spleen, Type I, saline solution
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%