Merck
Alcohol and oral squamous cell carcinoma.
SADJ : journal of the South African Dental Association = tydskrif van die Suid-Afrikaanse Tandheelkundige Vereniging (2013-08-27)
L Feller, R Chandran, R A G Khammissa, R Meyerov, J Lemmer
ABSTRACT
Alcohol is a risk factor for oral squamous cell carcinoma. It enhances the permeability of the oral epithelium, acts as a solvent for tobacco carcinogens, induces basal-cell proliferation, and generates free radicals and acetaldehyde, which have the capacity to cause DNA damage. Alcohol-associated malnutrition and immune suppression may further promote carcinogenesis. However, acetaldehyde, the first metabolite of ethanol, is the critical agent by which prolonged and excessive consumption of alcoholic beverages increases the risk of oral squamous cell carcinoma. Alcohol also acts synergistically with the products of tobacco combustion in the pathogenesis of oral squamous cell carcinoma.
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology