Merck
  • Metabolic disposition and pharmacokinetics of [14C]-amprenavir, a human immunodeficiency virus type 1 (HIV-1) protease inhibitor, administered as a single oral dose to healthy male subjects.
Metabolic disposition and pharmacokinetics of [14C]-amprenavir, a human immunodeficiency virus type 1 (HIV-1) protease inhibitor, administered as a single oral dose to healthy male subjects.
Journal of clinical pharmacology (2001-04-18)
B M Sadler, G E Chittick, R E Polk, D Slain, T M Kerkering, S D Studenberg, Y Lou, K H Moore, J L Woolley, D S Stein
ABSTRACT
The objective of this study was to determine the metabolic profile, routes of elimination, and total recovery of amprenavir and its metabolites after a single oral dose of [14C]-amprenavir. Six healthy male subjects each received a single oral 630 mg dose of amprenavir containing 95.76 microCi of [14C]-amprenavir in this Phase I mass balance study. The metabolic disposition of amprenavir was determined through analyses of radiocarbon in whole blood, plasma, urine, and stool samples, collected for a period of 10 to 17 days postdosing. Cerebral spinal fluid (CSF) sampling was conducted on day 1. The ratio of unchanged amprenavir AUC0-->infinity to plasma radiocarbon was 27%, suggesting that most of the radiocarbon was metabolites. The median total recovery of the administered dose of radiocarbon was 89% (range: 66%-93%), with 75% (range: 56%-80%) recovered in the feces and 14% (range: 10%-17%) in the urine. Most of the recovered radiocarbon in the feces and urine was excreted within 240 and 48 hours postdose, respectively. Of the 75% of the radiocarbon dose recovered in the feces, 62% was identified as a metabolite resulting from dioxidation of the tetrahydrofuran ring (GW549445X) and 32% as a metabolite resulting from subsequent oxidation of the p-aniline sulfonate group (GW549444X). Unchanged amprenavir was below the limit of quantitation in feces and urine. Therefore, approximately 94% of the dose excreted in the feces was accounted for by these two metabolites. Concentrations of radiocarbon in the CSF were below the limit of quantitation in 5 of 6 subjects sampled. In summary, oral amprenavir is extensively metabolized in humans, with concentrations of unchanged drug below the limits of quantitation in urine and feces. The majority (75%) of administered radiocarbon was excreted in feces.
Product Description
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%