Merck
  • Trans-differentiation of prostatic stromal cells leads to decreased glycoprotein hormone alpha production.
Trans-differentiation of prostatic stromal cells leads to decreased glycoprotein hormone alpha production.
The Journal of clinical endocrinology and metabolism (2002-11-05)
Holger Rumpold, Katarina Mascher, Gerold Untergasser, Eugen Plas, Martin Hermann, Peter Berger
ABSTRACT
Age-related development of benign prostatic hyperplasia is an important health issue in developed countries. The histopathogenetic hallmark of this disease is the increase in relative and absolute numbers of smooth muscle cells (SMC). Glycoprotein hormone alpha-subunit (GPHalpha) is expressed in the human prostate, and, because of its structural similarities to other cystine knot growth factors, it has been considered to have growth regulatory functions of its own. Primary cell cultures allowing for selective cultivation of prostatic epithelial cells, fibroblasts, and SMC were established. Directed trans-differentiation and cellular homogeneity was pursued by confocal scanning laser microscopy with cell type-specific markers. GPHalpha production by these cells was assessed by immunofluorimetric assays. Its predominant source was young fibroblasts, whereas replicative senescent fibroblasts, SMC, and control fibroblasts from foreskin did not produce significant amounts. Functionally, GPHalpha reduced growth of stromal cells at concentrations of 10 and 100 nmol/liter as shown by cell viability assays. It is concluded that histogenetic reorganization over the adult lifetime, guided by endocrine factors like steroid hormones together with senescence of fibroblasts, leads to a decreased production of growth inhibitors, such as GPHalpha, favoring proliferation and the development of benign prostatic hyperplasia.
Product Description
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cystine, Wacker Chemie AG, ≥98.5%
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
SAFC
L-Cystine
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Cystine, European Pharmacopoeia (EP) Reference Standard