Merck
  • Amino acid concentrations in cerebrospinal fluid in children with acute lymphoblastic leukemia undergoing chemotherapy.
Amino acid concentrations in cerebrospinal fluid in children with acute lymphoblastic leukemia undergoing chemotherapy.
European journal of cancer (Oxford, England : 1990) (2005-05-25)
C T Peng, K H Wu, S J Lan, Jeffrey J P Tsai, F J Tsai, C H Tsai
ABSTRACT
Cerebrospinal fluid (CSF) amino acid concentrations were measured in 45 children with acute lymphoblastic leukemia (ALL). Central nervous system (CNS) disease was absent in 34 and present in 11 (Groups L and M, respectively) at diagnosis. Thirty-two otherwise healthy children with febrile convulsions were studied for comparison. Results from this study show that glutamine levels at Day 0 were significantly higher in patients than in controls. Patients in Group M had elevated glutamine levels compared to Group L. In comparison, at Day 14, concentrations of glutamine and asparagine decreased, while glutamic acid amounts increased significantly in Group L. Glutamine levels fell at Day 42 in Group M, which may have resulted from more intensive treatment. From this study we hypothesise that higher baseline glutamine levels are indicative of a greater risk for CNS leukemia. Large-scale prospective trials are required to confirm increased baseline CSF glutamine levels in ALL patients, to identify glutamine as a marker for CNS disease and to clarify underlying mechanisms regulating glutamine in ALL.
Product Description
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
L-Tyrosine, FG
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
SAFC
Glycine
SAFC
L-Tyrosine
SAFC
L-Threonine
SAFC
L-Valine
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)