Merck
Transport of esterified retinol in fasting human blood.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition (1986-01-01)
R Schindler, A Klopp
ABSTRACT
The transport of retinyl palmitate in postabsorptive human blood has been investigated. For this purpose, lipoproteins were isolated from fasting serum of normal subjects as well as of patients with various diseases. The serum retinyl palmitate values of these individuals ranged from 2.0-30.3 micrograms/100 ml, with a mean value of 7.6 micrograms/100 ml. In the 14 human subjects investigated, 68.1% of the recovered retinyl palmitate was associated with the VLDL fraction and 28.9% with the LDL fraction. Nevertheless, the individual pattern of retinyl palmitate distribution among VLDL and LDL differed considerably within this group. No esterified retinol, however, was detected in the HDL as well as in the nonlipoprotein fraction (d greater than 1.21 g/ml) of human serum. It is concluded that the transport of esterified retinol in fasting blood is not restricted to chylomicron remnants. Nevertheless, in humans the serum transport of retinyl palmitate is preferentially mediated by lipoproteins of the VLDL fraction.
Sigma-Aldrich
Retinyl palmitate, potency: ≥1,700,000 USP units per g
Sigma-Aldrich
Retinyl palmitate, Type IV, ~1,800,000 USP units/g, oil
Supelco
Retinyl Palmitate (Vitamin A Palmitate), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Retinol palmitate, analytical standard