Merck
Quantitative analysis of catechol and 4-methylcatechol in human urine.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (1982-10-01)
S G Carmella, E J La Voie, S S Hecht
ABSTRACT
A method was developed for the quantitative analysis of catechol and 4-methylcatechol in human urine. [U-14C]Catechol was used as in internal standard. Urine was treated with beta-glucuronidase and sulphatase, acidified and extracted with ether. The ether extract was silylated and analysed by glass capillary gas chromatography. Catechol and 4-methylcatechol occurred in urine primarily as conjugates. Levels of catechol and 4-methylcatechol in the urine of nonsmokers on unrestricted diets were 10 +/- 7.3 (mean +/- 1 SD) and 3.4 +/- 2.3 mg/24 hr, respectively. Nonsmokers on uniform restricted diets, in which the intake of plant-derived products was limited, excreted 4.4 +/- 1.2 mg catechol and 8.1 +/- 1.7 mg 4-methylcatechol/24 hr. Smokers on the same restricted diet excreted 6.8 +/- 3.0 mg catechol and 6.1 +/- 2.6 mg 4-methylcatechol/24 hr. These results indicate that diet is a major factor in determining urinary catechol levels and that the contribution of smoking is comparatively small. Catechol and 4-methylcatechol appear to have different dietary precursors.
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
4-Methylcatechol, ≥95%
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
trans,trans-Muconic acid, 95%
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Pyrocatechol, ≥99%
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%