Merck
Serum cobalamin and methylmalonic acid in Alzheimer dementia.
Acta neurologica Scandinavica (1993-06-01)
M O Kristensen, N C Gulmann, J E Christensen, K Ostergaard, K Rasmussen
ABSTRACT
The cobalamin status was evaluated in Alzheimer dementia (n = 26), other dementias (n = 24), various gerontopsychiatric disorders (n = 25), and in neuro-psychiatrically healthy controls (n = 20). Supplementing serum cobalamin we measured methylmalonic acid (MMA), a metabolite accumulating early in cobalamin deficiency. Subnormal cobalamin and/or clearly elevated MMA concentrations were found in 11 cases: 7 Alzheimer patients (27%), 2 with other dementias (8%), one psychiatric patient (4%), and one control (5%). None presented the typical neurologic features of cobalamin deficiency and macrocytosis was found in only one. The mean cobalamin concentration was significantly lower in Alzheimer patients (179 +/- 18 pmol/l) than in the age-matched controls (256 +/- 23 pmol/l) (p = 0.013) and the other patient groups. Correspondingly, the mean MMA level was higher in the Alzheimer group (0.480 +/- 0.062 mumol/l) than in any other diagnostic group (controls: 0.347 +/- 0.040 mumol/l). Comparing the Alzheimer group to the other groups as a whole, the elevation was significant (p = 0.0097). Our findings indicate that Alzheimer patients are particularly prone to cobalamin deficiency, and even subtle biochemical signs of deficiency seem to justify treatment.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Mannitol, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis