- Interlayer for modified cathode in highly efficient inverted ITO-free organic solar cells.
Interlayer for modified cathode in highly efficient inverted ITO-free organic solar cells.
Advanced materials (Deerfield Beach, Fla.) (2012-01-18)
Zheng Tang, L Mattias Andersson, Zandra George, Koen Vandewal, Kristofer Tvingstedt, Patrik Heriksson, Renee Kroon, Mats R Andersson, Olle Inganäs
PMID22250035
ABSTRACT
Inverted polymer solar cells with a bottom metal cathode modified by a conjugated polymer interlayer show considerable improvement of photocurrent and fill factor, which is due to hole blocking at the interlayer, and a modified surface energy which affects the nanostructure in the TQ1/[70]PCBM blend.