Merck
  • Modified split-cast technique: a new, timesaving clinical remount technique.
Modified split-cast technique: a new, timesaving clinical remount technique.
Journal of prosthodontics : official journal of the American College of Prosthodontists (2010-06-29)
Fang-Chun Liu, Kwing-Chi Luk, Piin-Chau Suen, Tzong-shiun Tsai, Yen-Chen Ku
ABSTRACT
Interocclusal discrepancies can be eliminated by a clinical remount procedure, but most practitioners avoid it because of the time involved. This article introduces a new timesaving method, the modified split-cast technique. It uses a semi-adjustable articulator, tin foil as plaster separator, and an addition-type, silicone bite-registration material. The technician does most of the remounting procedures before the denture delivery appointment, so the dentist spends very little time chairside to complete the clinical remount procedure. Compared with the conventional and two other remounting techniques, the new technique is faster and easier to manipulate.
Product Description
Tin, foil, not light tested, 150x150mm, thickness 0.006mm, 97.4%
Tin, foil, not light tested, 150x150mm, thickness 0.007mm, 97.4%
Tin, foil, light tested, 50x50mm, thickness 0.008mm, 99.75%
Tin, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Tin, foil, 50x50mm, thickness 0.65mm, as rolled, 99.99+%
Tin, foil, 300x300mm, thickness 0.15mm, as rolled, 98.8%
Tin, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99+%
Tin, foil, 0.1m coil, thickness 0.06mm, coil width 73mm, as rolled, 99.99+%
Tin, foil, 50x50mm, thickness 0.15mm, as rolled, 98.8%
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99+%
Tin, foil, 50x50mm, thickness 0.75mm, as rolled, 99.99+%
Tin, foil, 25x25mm, thickness 0.1mm, as rolled, 98.8%
Tin, foil, 0.5m coil, thickness 0.5mm, as rolled, 98.8%
Tin, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Tin, foil, 0.2m coil, thickness 0.06mm, coil width 73mm, as rolled, 99.99+%
Tin, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 99.999%
Tin, foil, 10m coil, thickness 0.084mm, coil width 41mm, as rolled, 99.95%
Tin, foil, 100x100mm, thickness 0.65mm, as rolled, 99.99+%
Tin, foil, 0.5m coil, thickness 0.05mm, as rolled, 98.8%
Tin, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99+%
Tin, foil, 300x300mm, thickness 0.20mm, as rolled, 98.8%
Tin, foil, 150x150mm, thickness 0.15mm, as rolled, 98.8%
Tin, foil, 150x150mm, thickness 0.125mm, as rolled, 98.8%
Tin, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95%
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 98.8%
Tin, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Tin, foil, 50x50mm, thickness 0.1mm, as rolled, 98.8%
Tin, foil, 25x25mm, thickness 0.75mm, as rolled, 99.99+%
Tin, foil, 10m coil, thickness 0.007mm, 97.4%