- Coverage of denture teeth with tin foil for radiographic guide fabrication.
Coverage of denture teeth with tin foil for radiographic guide fabrication.
Journal of prosthodontics : official journal of the American College of Prosthodontists (2013-07-31)
Abdulmohsin Alhashim, Edward Robert Chesla
PMID23890162
ABSTRACT
A novel technique using the patient's existing complete dentures as a radiographic guide for diagnosis and treatment planning in implant dentistry is presented. Tin foil is used to cover the denture teeth before the radiographic scan is performed. Advantages of the described technique include its cost-effectiveness, simplicity, efficiency, and lack of need to modify or duplicate the patient's existing dentures. A disadvantage of the technique is that it serves only as a radiographic guide.
Product Description
Tin, foil, 100x100mm, thickness 0.15mm, as rolled, 98.8%
Tin, foil, 100x100mm, thickness 0.75mm, as rolled, 99.99+%
Tin, foil, 100x100mm, thickness 1.0mm, as rolled, 98.8%
Tin, foil, 100x100mm, thickness 0.1mm, as rolled, 98.8%
Tin, foil, 150x150mm, thickness 2.0mm, as rolled, 99.99+%
Tin, foil, 25x25mm, thickness 0.125mm, as rolled, 98.8%
Tin, foil, 100x100mm, thickness 1.0mm, as rolled, 99.999%
Tin, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Tin, foil, 150x150mm, thickness 0.25mm, as rolled, 98.8%
Tin, foil, 100x100mm, thickness 1.5mm, as rolled, 99.99+%
Tin, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Tin, foil, 50x50mm, thickness 0.65mm, as rolled, 99.99+%
Tin, foil, 300x300mm, thickness 0.15mm, as rolled, 98.8%
Tin, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99+%
Tin, foil, 0.1m coil, thickness 0.06mm, coil width 73mm, as rolled, 99.99+%
Tin, foil, 50x50mm, thickness 0.15mm, as rolled, 98.8%
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99+%
Tin, foil, 50x50mm, thickness 0.75mm, as rolled, 99.99+%
Tin, foil, 25x25mm, thickness 0.1mm, as rolled, 98.8%
Tin, foil, 0.5m coil, thickness 0.5mm, as rolled, 98.8%
Tin, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Tin, foil, 0.2m coil, thickness 0.06mm, coil width 73mm, as rolled, 99.99+%
Tin, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 99.999%
Tin, foil, 10m coil, thickness 0.084mm, coil width 41mm, as rolled, 99.95%
Tin, foil, 100x100mm, thickness 0.65mm, as rolled, 99.99+%
Tin, foil, 4mm disks, thickness 0.02mm, 99.75%
Tin, foil, 25mm disks, thickness 0.5mm, as rolled, 99.99+%
Tin, foil, 25mm disks, thickness 0.01mm, 97.4%
Tin, foil, 50mm disks, thickness 0.030mm, as rolled, 97.4%