Merck
  • Processing factors affecting the clarity of a rapid-curing clear acrylic resin.
Processing factors affecting the clarity of a rapid-curing clear acrylic resin.
Journal of oral rehabilitation (1979-10-01)
S B Keng, D W Cruickshanks-Boyd, E H Davies
PMID291704
ABSTRACT
The difficulty in repeatedly producing unblemished, clear acrylic resin in the dental laboratory has hindered its wider use, despite its many advantages over coloured material. Recently, rapid-cure dental acrylics have been introduced, which are available in both clear and coloured forms. This investigation examined various factors which may influence the production of unblemished, rapid-curing, clear acrylic resin. Utilizing a quantitative assessment of clarity, the most important factor influencing the clarity of the resin is shown to be the choice of separating medium. Tin-foil produces extremely high clarity, but alginate mould separator causes surface blanching. However, this surface blanching can be removed by polishing. Porosity, caused by too rapid curing, and stone model dryness are of only secondary importance. Possible water contamination of the monomer liquid due to accidental exposure only affects clarity at very high levels of contamination.
Product Description
Tin, foil, not light tested, 150x150mm, thickness 0.007mm, 97.4%
Tin, foil, not light tested, 150x150mm, thickness 0.006mm, 97.4%
Tin, foil, light tested, 50x50mm, thickness 0.008mm, 99.75%
Tin, foil, not light tested, 150x150mm, thickness 0.009mm, 97.4%
Tin, foil, not light tested, 25x25mm, thickness 0.006mm, 99.75%
Tin, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99+%
Tin, foil, 50x50mm, thickness 0.15mm, as rolled, 98.8%
Tin, foil, not light tested, 25x25mm, thickness 0.004mm, 99.75%
Tin, foil, 15mm disks, thickness 0.0125mm, 99.75%
Tin, foil, 2m coil, thickness 0.009mm, 97.4%
Tin, foil, 15mm disks, thickness 0.008mm, 99.75%
Tin, foil, 10mm disks, thickness 0.1mm, as rolled, 99.95%
Tin, foil, 10mm disks, thickness 1.0mm, as rolled, 98.8%
Tin, foil, 10mm disks, thickness 0.01mm, 97.4%
Tin, foil, 15mm disks, thickness 0.15mm, as rolled, 98.8%
Tin, foil, 1m coil, thickness 0.109mm, coil width 41mm, as rolled, 99.95%
Tin, foil, 15mm disks, thickness 0.5mm, as rolled, 99.99+%
Tin, foil, 1m coil, thickness 0.075mm, as rolled, 98.8%
Tin, foil, 15mm disks, thickness 0.025mm, as rolled, 99.99+%
Tin, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Tin, foil, 10mm disks, thickness 0.025mm, as rolled, 98.8%
Tin, foil, 10mm disks, thickness 1.0mm, as rolled, 99.99+%
Tin, foil, 15mm disks, thickness 0.15mm, as rolled, 99.99+%
Tin, foil, 2m coil, thickness 0.072mm, coil width 41mm, as rolled, 99.95%
Tin, foil, 2m coil, thickness 0.0125mm, 97.4%
Tin, foil, 15mm disks, thickness 0.009mm, 97.4%
Tin, foil, 25mm disks, thickness 0.5mm, as rolled, 98.8%
Tin, foil, 25mm disks, thickness 0.015mm, 99.75%
Tin, foil, 2m coil, thickness 0.084mm, coil width 41mm, as rolled, 99.95%
Tin, foil, 10mm disks, thickness 0.01mm, 99.75%