- Evaluation of the repair of mechanical furcation perforations using amalgam, gutta-percha, or indium foil.
Evaluation of the repair of mechanical furcation perforations using amalgam, gutta-percha, or indium foil.
Journal of endodontics (1986-06-01)
R Aguirre, M E elDeeb, M E elDeeb
PMID3461111
Product Description
Indium, foil, 6mm disks, thickness 0.05mm, as rolled, 100%
Indium, foil, 4mm disks, thickness 0.75mm, as rolled, 100%
Indium, foil, 6mm disks, thickness 0.125mm, as rolled, 100%
Indium, foil, 6mm disks, thickness 0.075mm, as rolled, 100%
Indium, foil, 8mm disks, thickness 0.03mm, 99.8%
Indium, foil, not light tested, 100x100mm, thickness 0.03mm, 99.8%
Indium, foil, not light tested, 100x100mm, thickness 0.006mm, temporary acrylic support, 99.8%
Indium, foil, 8mm disks, thickness 0.015mm, 99.8%
Indium, foil, not light tested, 100x100mm, thickness 0.015mm, 99.8%
Indium, foil, not light tested, 100x100mm, thickness 0.04mm, 99.8%
Indium, foil, 8mm disks, thickness 0.04mm, 99.8%
Indium, foil, light tested, 100x100mm, thickness 0.075mm, as rolled, 100%
Indium, foil, 8mm disks, thickness 0.05mm, as rolled, 100%
Indium, foil, not light tested, 25x25mm, thickness 0.0025mm, temporary acrylic support, 99.8%
Indium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 100%
Indium, foil, 8mm disks, thickness 0.075mm, as rolled, 100%
Indium, foil, 6mm disks, thickness 0.5mm, as rolled, 100%
Indium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 100%
Indium, foil, not light tested, 100x100mm, thickness 0.01mm, 99.8%
Indium, foil, not light tested, 100x100mm, thickness 0.009mm, temporary acrylic support, 99.8%
Indium, foil, not light tested, 100x100mm, thickness 0.02mm, 99.8%
Indium, foil, 6mm disks, thickness 0.75mm, as rolled, 100%
Indium, foil, not light tested, 25x25mm, thickness 0.003mm, temporary acrylic support, 99.8%
Indium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 100%
Indium, foil, not light tested, 100x100mm, thickness 0.008mm, temporary acrylic support, 99.8%
Indium, foil, 8mm disks, thickness 0.0125mm, temporary acrylic support, 99.8%
Indium, foil, not light tested, 100x100mm, thickness 0.0125mm, temporary acrylic support, 99.8%
Indium, foil, 8mm disks, thickness 0.1mm, as rolled, 100%
Indium, foil, not light tested, 25x25mm, thickness 0.004mm, temporary acrylic support, 99.8%
Indium, foil, not light tested, 50x50mm, thickness 0.0025mm, temporary acrylic support, 99.8%